Läs mer
3M™ Tora™ Safety Glasses
Handla allt 3M Produkter
:
Lens Tint: Bronze
Beskrivning
- Safety glasses with a wraparound lens design offering excellent peripheral vision and lateral side protection
- Anti-scratch and anti-fog coating for increased durability and improved vision
- Extremely lightweight at only 22 g
- Adjustable neck cord for increased comfort and convenient storage
Specifikationer
Specifikationer
| Certifikat och efterlevnad | CE Approved, EN 166:2001 |
| Linsbeläggning | Anti-fog, Scratch-resistant |
| Linsfärg | Bronze |
| Produkttyp | Safety glasses |
| Linsmaterial | Polycarbonate |