Läs mer
Torsemide Carboxylic Acid, TRC
Produktkod. 30450763
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30450763 50 mg
1 options
Produktkod. 30450763 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT54875550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Torsemide Carboxylic Acid
CAS 113844-99-8
Molekylformel C16H18N4O5S
Kvantitet 50 mg
Synonym 3-[[3-[[[[(1-Methylethyl)amino]carbonyl]amino]sulfonyl]-4-pyridinyl]amino]-benzoic Acid, 3-((3-(N-(Isopropylcarbamoyl)sulfamoyl)pyridin-4-yl)amino)benzoic Acid
InChI formel InChI=1S/C16H18N4O5S/c1-10(2)18-16(23)20-26(24,25)14-9-17-7-6-13(14)19-12-5-3-4-11(8-12)15(21)22/h3-10H,1-2H3,(H,17,19)(H,21,22)(H2,18,20,23)
LEDER CC(C)NC(NS(C1=CN=CC=C1NC2=CC=CC(C(O)=O)=C2)(=O)=O)=O
Molekylvikt (g/mol) 378.4
Formel vikt 378.0998