Läs mer
Total Base Number Standard, ASTM D2896 (nominal value: 10.01 mg KOH/g), 1 x 50g, Paragon Scientific™
Produktkod. 16070440
Change view
Kvantitet:
1 x 50 g
Förpackningsstorlek:
Paket med 3
Produktkod. Kvantitet unitSize
16070440 1 x 50 g Paket med 3
1 options
Produktkod. 16070440 Leverantör Paragon Scientific Leverantörsnummer TBN10/3
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Specifically manufactured for the verification of analytical instruments used to determine base number by potentiometric titration. Each standard is certified in strict accordance with international test method protocols, ASTM D2896 / IP 276.
  • Base Number (TBN) tested and certified in accordance with ASTM D2896 / IP 276
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
  • Typically used in, but not limited to, the analysis of used oils and lubricants
  • Supplied as single volume packs or as triple packs
  • Supplied in tamper-evident glass packaging, providing assurance of sample integrity
  • Manufactured in the United Kingdom
  • 24 month shelf life
Specifikationer
Kvantitet 1 x 50 g
Nominal values quoted. Certified Values will vary from batch to batch.