missing translation for 'onlineSavingsMsg'
Learn More

TP-0903, TRC

Product Code. 30453692
Change view
Click to view available options
Kvantitet:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30453692 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453692 Supplier Toronto Research Chemicals Supplier No. TRCT7003002.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-[(5-Chloro-2-[(4-[(4-methyl-1-piperazinyl)methyl]phenyl)amino]-4-pyrimidinyl)amino]-N,N-dimethyl-benzenesulfonamide
CAS 1341200-45-0
Molekylformel C24H30ClN7O2S
Kvantitet 2.5 mg
Synonym TP-0903
InChI formel InChI=1S/C24H30ClN7O2S/c1-30(2)35(33,34)22-7-5-4-6-21(22)28-23-20(25)16-26-24(29-23)27-19-10-8-18(9-11-19)17-32-14-12-31(3)13-15-32/h4-11,16H,12-15,17H2,1-3H3,(H2,26,27,28,29)
LEDER CN(C)S(=O)(=O)c1ccccc1Nc2nc(Nc3ccc(CN4CCN(C)CC4)cc3)ncc2Cl
IUPAC-namn 2-[[5-chloro-2-[4-[(4-methylpiperazin-1-yl)methyl]anilino]pyrimidin-4-yl]amino]-N,N-dimethylbenzenesulfonamide
Molekylvikt (g/mol) 516.06
Formel vikt 515.187
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.