missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ TPI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599973
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Antibody detects endogenous levels of total TPI1.
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
Specifikationer
| TPI1 | |
| Polyclonal | |
| Unconjugated | |
| Tpi1 | |
| AI255506; epididymis secretory protein Li 49; HEL-S-49; Methylglyoxal synthase; MGC88108; TIM; Tpi; TPI1; Tpi-1; TPID; triosephosphate isomerase; triose-phosphate isomerase; triosephosphate isomerase 1; triose-phosphate isomerase TPI1; unnamed protein product; YD9609.05C; YDR050C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21991, 24849, 7167 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17751, P48500, P60174 | |
| Tpi1 | |
| A synthesized peptide derived from human TPI1(Accession P60174), corresponding to amino acid residues T254-Q286. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering