missing translation for 'onlineSavingsMsg'
Learn More

TPM4 Recombinant Protein, Abnova

Brand:  Abnova H00007171-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16181532

  • 5030.41 SEK / 25µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH37576
Solution
7171
TPM4 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TPM4
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
53.02
8
Glutathione Sepharose 4 Fast Flow
25 ug
MAGLNSLEAVKRKIQALQQQADEAEDRAQGLQRELDGERERREKAEGDVAALNRRIQLVEEELDRAQERLATALQKLEEAEKAADESERGMKVIENRAMKDEEKMEIQEMQLKEAKHIAEEADRKYEEVARKLVILEGELERAEERAEVSELKCGDLEEELKNVTNNLKSLEAASEKYSEKEDKYEEEIKLLSDKLKEAETRAEFAERTVAKLEKTIDDLEEKLAQAKEENVGLHQTLDQTLNELNCI
TPM4
Human
Yes
Videos
SDS
Documents
Certificates