Learn More
TraceGOLD™ TG-17SilMS GC Columns
Optimize separation when analyzing active environmental compounds with this low-bleed GC column with superior inertness.
Brand:  Thermo Scientific 26072-1310
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15244533
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
Engineered for Superior Performance
  • Midpolarity phase similar to 50% phenyl methylpolysiloxane
  • High thermal stability — maximum temperatures up to 340° to 360°C
Applications
  • PAHs
  • Pesticides and herbicides
  • Phthalate esters
  • Triglycerides
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • A low-bleed, stable column produces consistent results
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes
  • Minimizing column bleed extends column lifetime, leading to higher productivity
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility
Recommended For:
  • Herbicides
  • PAHs
  • Pesticides
  • Phthalate esters
Alternative Uses
  • Triglycerides
  • Ketones
  • Nitroaromatics
  • Plant sterols
  • Pyrethroids
Similar To:
  • DB-17ms
  • VF-17ms
  • CP-Sil 24 CB
  • ZB-50
  • BPX-50
Specifications
Specifications
G3
0.25µm
15m
0.32mm
Videos
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.