Learn More
Thermo Scientific™ TraceGOLD™ TG-SQC GC Columns
Product Code. 10157664 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
15 m
30 m
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10157664 30 m Each
10127904 15 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10157664 Supplier Thermo Scientific™ Supplier No. 260701420
Beräknad leverans: 30-11-2026
Ensure the quality of your Thermo Scientific instrument when installed on site.
Ensure the quality of your Thermo Scientific instrument when installed on site by using the Thermo Scientific™ TraceGOLD™ TG-SQC GC column. The TraceGOLD TG-SQC GC column is optimized for system qualification tests during initial installation, or during service and maintenance of an installed instrument. This column is recommended for benchmark testing only.
Applications:
System qualification tests (Thermo Scientific GC-MS)
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 30 m
Diameter (Metric) Inner 0.25 mm
Functionality/Interaction Type Benchmark Testing Only
Product Line TraceGOLD
Film Thickness 0.25 μm
Quantity Each
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.