Läs mer
TRADD Antibody (313203), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2658D
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TRADD Monoclonal antibody specifically detects TRADD in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifikationer
| TRADD | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 313203 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| MGC11078, TNFRSF1A-associated via death domainTNFR1-associated DEATH domain protein, tumor necrosis factor receptor type 1 associated death domain protein, tumor necrosis factor receptor type 1-associated DEATH domain protein, tumor necrosis factor receptor-1-associated protein | |
| E. coli-derived recombinant human TRADD, Met1-Ala312, Accession # Q15628 | |
| 0.1 mL | |
| Apoptosis, Cancer, Death Receptor Signaling Pathway, Tumor Suppressors | |
| 8717 | |
| Store at 4°C in the dark. | |
| IgG2a |