missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
TRAF-3 Antibody (PSH01-30), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33042
This item is not returnable.
View return policy
Description
TRAF-3 Monoclonal antibody specifically detects TRAF-3 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| TRAF-3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3 | |
| Recombinant protein within human TRAF-3 301-568/ 568. (Uniprot: Q13114) | |
| 100 μL | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| 7187 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| PSH01-30 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction