missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12705-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TRAM, or TRIF-related adaptor molecule, plays an essential role in the MyD88-independent signaling of TLR4.Specifications
TRAM1 | |
Polyclonal | |
Unconjugated | |
TRAM1 | |
PNAS 8, PRO1292, TRAM, TRAM1, TRAMP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23471, 312903, 72265 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15629, Q5XI41, Q91V04 | |
Tram1 | |
TRAM1 Fusion Protein Ag3384 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |