Learn More
Tramadol N-Oxide, Mikromol™
Product Code. 18083100
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18083100 100 mg 100mg
30050513 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18083100 Supplier Mikromol Supplier No. MM0007.11
 Restricted Item
Beräknad leverans: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tramadol N-Oxide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tramadol N-Oxide
CAS 147441-56-3
Analyte or Component Names Tramadol N-Oxide
Shipping Condition Room Temperature
Molecular Formula C16 H25 N O3
Certifications/Compliance ISO 9001
Synonym Tramadol N-Oxide
InChI Formula InChI=1S/C16H25NO3/c1-17(2,19)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)20-3/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3/t14-,16+/m1/s1
SMILES COc1cccc(c1)[C@@]2(O)CCCC[C@@H]2C[N+](C)(C)[O-]
IUPAC Name 1-[(1R,2R)-2-hydroxy-2-(3-methoxyphenyl)cyclohexyl]-N,N-dimethylmethanamine oxide
Molecular Weight (g/mol) 279.37
Quantity 100 mg
Formula Weight 279.1834 g/mol