Läs mer
Tramadol N-Oxide, Mikromol™
Produktkod. 30050513
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050513 25 mg
18083100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050513 Leverantör Mikromol Leverantörsnummer MM0007.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tramadol N-Oxide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tramadol N-Oxide
CAS 147441-56-3
Analyt- eller komponentnamn Tramadol N-Oxide
Frakt skick Room Temperature
Molekylformel C16 H25 N O3
Certifikat och efterlevnad ISO 9001
Synonym Tramadol N-Oxide
InChI formel InChI=1S/C16H25NO3/c1-17(2,19)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)20-3/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3/t14-,16+/m1/s1
LEDER COc1cccc(c1)[C@@]2(O)CCCC[C@@H]2C[N+](C)(C)[O-]
IUPAC-namn 1-[(1R,2R)-2-hydroxy-2-(3-methoxyphenyl)cyclohexyl]-N,N-dimethylmethanamine oxide
Molekylvikt (g/mol) 279.37
Kvantitet 25 mg
Formel vikt 279.1834 g/mol