missing translation for 'onlineSavingsMsg'
Learn More

Tranexamic acid, MedChemExpress

MedChemExpress Tranexamic acid (Transamin) is an antifibrinolytic for blocking lysine-binding sites of plasmin and elastase-derived plasminogen fragments with IC50 of 5 mM.

964.76 SEK - 1353.77 SEK

Specifications

Chemical Name or Material Tranexamic acid
Physical Form Solid
CAS 1197-18-8
Color White
Purity Grade Notes Research
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16470788
Encompass_Preferred
View Documents
Medchem Express
HY-B0149
5 g
964.76 SEK
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16428368
Encompass_Preferred
View Documents
Medchem Express
HY-B0149
10 g
1353.77 SEK
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tranexamic acid (Transamin) is an antifibrinolytic for blocking lysine-binding sites of plasmin and elastase-derived plasminogen fragments with IC50 of 5 mM.
Specifications

Specifications

Tranexamic acid
1197-18-8
Research
COVID-19-immunoregulation
O=C([C@H]1CC[C@H](CN)CC1)O
157.21
Research
Solid
White
C8H15NO2
H2O : 110 mg/mL (699.70 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
157.21
98.0%
Videos
Safety and Handling

Safety and Handling

H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.