Läs mer
trans-1,2-Dichlorocyclohexane, TRC
Produktkod. 30449690
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449690 50 mg
30448951 500 mg
30456686 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30449690 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT70513050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 822-86-6
Molekylformel C6H10Cl2
Molekylvikt (g/mol) 153.05
IUPAC-namn (1R,2R)-1,2-dichlorocyclohexane
LEDER Cl[C@@H]1CCCC[C@H]1Cl
Specifikationer
Kemiskt namn eller material trans-1,2-Dichlorocyclohexane
Kvantitet 50 mg
InChI formel InChI=1S/C6H10Cl2/c7-5-3-1-2-4-6(5)8/h5-6H,1-4H2/t5-,6-/m1/s1
Formel vikt 152.016