missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2,3-Epoxybutane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.
Solubility
Soluble in water(95g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.
Solubility
Soluble in water(95g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications
Density | 0.804 |
Boiling Point | 54°C to 55°C |
Flash Point | −26°C (−14°F) |
Odor | Recognizable |
Refractive Index | 1.373 |
Quantity | 1 g |
UN Number | UN3271 |
Beilstein | 79772 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water(95g/L). |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'