Läs mer
trans-2-Pinanol, TRC
Produktkod. 30419434
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419434 10 mg
30426730 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30419434 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP45862310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4948-29-2
Molekylformel C10H18O
Molekylvikt (g/mol) 154.25
Synonym rel-(1R,2R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptan-2-ol, NSC 2326, Pinan-2beta-ol
IUPAC-namn (1S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-4-ol
LEDER C[C@@]1(O)CC[C@H]2C[C@@H]1C2(C)C
Specifikationer
Kemiskt namn eller material trans-2-Pinanol
Anteckningar om renhetsgrad GC
Kvantitet 10 mg
InChI formel InChI=1S/C10H18O/c1-9(2)7-4-5-10(3,11)8(9)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10+/m0/s1
Formel vikt 154.1358
Procent renhet >95%