Läs mer
trans-3-Hexenedioic Acid, TRC
Produktkod. 30406981
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30406981 1 g
1 options
Produktkod. 30406981 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2949051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans-3-Hexenedioic Acid
CAS 4436-74-2
Molekylformel C6H8O4
Kvantitet 1 g
Synonym 3-Hexenedioic Acid, beta-Hydromuconic Acid, 2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid
InChI formel InChI=1S/C6H8O4/c7-5(8)3-1-2-4-6(9)10/h1-2H,3-4H2,(H,7,8)(H,9,10)/b2-1+
LEDER OC(=O)C\C=C\CC(=O)O
IUPAC-namn (E)-hex-3-enedioic acid
Molekylvikt (g/mol) 144.13
Formel vikt 144.0423