Läs mer
trans-3'-Hydroxy Cotinine Acetate, TRC
Produktkod. 30407367
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30407367 25 mg
1 options
Produktkod. 30407367 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH92450825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3'R,5'S)-3'-Hydroxycotinine Acetate
CAS 111034-55-0
Molekylformel C12 H14 N2 O3
Kvantitet 25 mg
Synonym 2-Pyrrolidinone, 3-(acetyloxy)-1-methyl-5-(3-pyridinyl)-, (3R-trans)- (9CI), Hydroxycotinine acetate, trans-3'-Hydroxycotinine acetate, trans-3'-Hydroxy cotinine acetate, (3'R,5'S)-3'-Hydroxycotinine acetate, (3R,5S)-3-Acetoxy-1-methyl-5-(3-pyridinyl)-2-pyrrolidinone
InChI formel InChI=1S/C12H14N2O3/c1-8(15)17-11-6-10(14(2)12(11)16)9-4-3-5-13-7-9/h3-5,7,10-11H,6H2,1-2H3/t10-,11+/m1/s1
LEDER CN1[C@H](C[C@H](OC(=O)C)C1=O)c2cccnc2
IUPAC-namn [(3S,5R)-1-methyl-2-oxo-5-pyridin-3-ylpyrrolidin-3-yl] acetate
Molekylvikt (g/mol) 234.25
Formel vikt 234.1004