Läs mer
trans-3'-Hydroxy Cotinine N-Beta-D-Glucuronide, TRC
Produktkod. 30132850
Change view
Kvantitet:
1 mg
2 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132850 2 mg
30138139 5 mg
30008932 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132850 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9245302MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 146275-18-5
Molekylformel C16 H20 N2 O8
Molekylvikt (g/mol) 368.34
Synonym Pyridinium, 1-β-D-glucopyranuronosyl-3-[(2S,4R)-4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl]-, inner salt (9CI, ACI), Pyridinium, 1-β-D-glucopyranuronosyl-3-(4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl)-, inner salt, (2S-trans)- (ZCI), trans-3'-Hydroxycotinine-N-glucuronide, trans-3'-Hydroxy Cotinine N-β-D-Glucuronide, 1-β-D-Glucopyranuronosyl-3-[(2S,4R)-4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl]pyridinium inner salt, (3R,5S)-3-Hydroxy-1-methyl-5-(3'-pyridinyl)-2-pyrrolidinone N'-β-D-Glucuronide, trans-3'-Hydroxycotinine N-β-D-Glucuronide
IUPAC-namn (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[(3R,5S)-1-methyl-2-oxo-5-pyridin-3-ylpyrrolidin-3-yl]oxyoxane-2-carboxylic acid
LEDER CN1[C@@H](C[C@@H](O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)C1=O)c3cccnc3
Specifikationer
Kemiskt namn eller material trans-3'-Hydroxycotinine-N-beta-D-glucuronide
Kvantitet 2 mg
InChI formel InChI=1S/C16H20N2O8/c1-18-8(7-3-2-4-17-6-7)5-9(14(18)22)25-16-12(21)10(19)11(20)13(26-16)15(23)24/h2-4,6,8-13,16,19-21H,5H2,1H3,(H,23,24)/t8-,9+,10-,11-,12+,13-,16+/m0/s1
Formel vikt 368.122