Läs mer
trans-4-(6,8-Dibromoquinazolin-3(4H)-yl)cyclohexanol Hydrochloride, Mikromol™
Produktkod. 30049570
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049570 25 mg
17903446 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049570 Leverantör Mikromol Leverantörsnummer MM0005.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
trans-4-(6,8-Dibromoquinazolin-3(4H)-yl)cyclohexanol Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans-4-(6,8-Dibromoquinazolin-3(4H)-yl)cyclohexanol Hydrochloride
CAS 2356152-56-0
Analyt- eller komponentnamn trans-4-(6,8-Dibromoquinazolin-3(4H)-yl)cyclohexanol Hydrochloride
Frakt skick Room Temperature
Molekylformel C14 H16 Br2 N2 O . Cl H
Certifikat och efterlevnad ISO 9001
Synonym trans-4-(6,8-Dibromoquinazolin-3(4H)-yl)cyclohexanol Hydrochloride
InChI formel InChI=1S/C14H16Br2N2O.ClH/c15-10-5-9-7-18(8-17-14(9)13(16)6-10)11-1-3-12(19)4-2-11;/h5-6,8,11-12,19H,1-4,7H2;1H/t11-,12-;
LEDER Cl.O[C@@H]1CC[C@H](CC1)N2Cc3cc(Br)cc(Br)c3N=C2
IUPAC-namn 4-(6,8-dibromo-4H-quinazolin-3-yl)cyclohexan-1-ol;hydrochloride
Molekylvikt (g/mol) 424.56
Kvantitet 25 mg
Formel vikt 421.9396 g/mol