Learn More
trans-4-(Boc-aminomethyl)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Used as pharmaecutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaecutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Used as pharmaecutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Solubility Information | Slightly soluble in water. |
Formula Weight | 242.35 |
Percent Purity | 97% |
Quantity | 250 mg |
Chemical Name or Material | trans-4-(Boc-aminomethyl)cyclohexanemethylamine |