missing translation for 'onlineSavingsMsg'
Läs mer

trans-4-Chlorocinnamic Acid, TRC

Produktkod. 30368385
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368385 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30368385 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36508010G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material trans-4-Chlorocinnamic Acid
CAS 940-62-5
Molekylformel C9H7ClO2
Kvantitet 10 g
Synonym (2E)-3-(4-Chlorophenyl)-2-propenoic Acid, NSC 52172
InChI formel InChI=1S/C9H7ClO2/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6H,(H,11,12)/b6-3+
LEDER OC(=O)\C=C\c1ccc(Cl)cc1
IUPAC-namn (E)-3-(4-chlorophenyl)prop-2-enoic acid
Molekylvikt (g/mol) 182.6
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.