Learn More
trans-4-Decenal, 96%, may cont. ca 7% cis-isomer
Product Code. 11377565
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377565 25 g 25g
11367565 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11377565 Supplier Thermo Scientific Alfa Aesar Supplier No. L05715.14
 Restricted Item
I lager
Add to basket
CAS: 65405-70-1 | C10H18O | 154.253 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65405-70-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00065447
InChI Key CWRKZMLUDFBPAO-VOTSOKGWSA-N
Synonym trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264
PubChem CID 5702654
IUPAC Name (E)-dec-4-enal
SMILES CCCCCC=CCCC=O
Specifications
Density 0.842
Boiling Point 80°C (4 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.445
Quantity 25 g
Beilstein 4230058
Formula Weight 154.25
Percent Purity 96%
Assay may cont. ≈7% cis-isomer
Chemical Name or Material trans-4-Decenal