Läs mer
trans-7,8-Dihydroxy-7,8-dihydrobenzo[a]pyrene, TRC
Produktkod. 30010341
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010341 2.5 mg 2.5mg
30006672 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010341 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4522802.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57404-88-3
Molekylformel C20 H14 O2
Molekylvikt (g/mol) 286.32
Synonym Benzo[a]pyrene-7,8-diol, 7,8-dihydro-, (7R,8R)-rel-, Benzo[a]pyrene-7,8-diol, 7,8-dihydro-, trans-, rel-(7R,8R)-7,8-Dihydrobenzo[a]pyrene-7,8-diol, (±)-trans-7,8-Dihydro-7,8-dihydroxybenzo[a]pyrene, (±)-trans-7,8-Dihydroxy-7,8-dihydrobenzo[a]pyrene, Benzo[a]pyrene-trans-7,8-dihydrodiol, trans-7,8-Dihydro-7,8-dihydroxybenzo[a]pyrene, trans-7,8-Dihydrobenzo[a]pyrene-7,8-diol, trans-7,8-Dihydroxy-7,8-dihydrobenzo[a]pyrene, trans-Benzo[a]pyrene-7,8-dihydrodiol
IUPAC-namn (7S,8S)-7,8-dihydrobenzo[a]pyrene-7,8-diol
LEDER O[C@H]1C=Cc2c(cc3ccc4cccc5ccc2c3c45)[C@@H]1O
Specifikationer
Kemiskt namn eller material (±)-trans-7,8-Dihydroxy-7,8-dihydrobenzo[a]pyrene
Kvantitet 2.5 mg
InChI formel InChI=1S/C20H14O2/c21-17-9-8-14-15-7-6-12-3-1-2-11-4-5-13(19(15)18(11)12)10-16(14)20(17)22/h1-10,17,20-22H/t17-,20-/m0/s1
Formel vikt 286.0994