Läs mer
trans-Amorolfine, TRC
Produktkod. 30133498
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133498 50 mg
30011285 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133498 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63416550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2134097-34-8
Molekylformel C21 H35 N O
Molekylvikt (g/mol) 317.51
Synonym Morpholine, 4-[3-[4-(1,1-dimethylpropyl)phenyl]-2-methylpropyl]-2,6-dimethyl-, (2R,6R)-rel-, (2RS,6RS)-2,6-Dimethyl-4-[2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl]morpholine, (2RS,6RS)-4-[3-[4-(1,1-Dimethylpropyl)phenyl]-2-methyl-propyl]-2,6-dimethyl-morpholine, trans-Amorolfine
IUPAC-namn (2S,6S)-2,6-dimethyl-4-[2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl]morpholine
LEDER CCC(C)(C)c1ccc(CC(C)CN2C[C@H](C)O[C@@H](C)C2)cc1
Specifikationer
Kemiskt namn eller material trans-Amorolfine
Kvantitet 50 mg
InChI formel InChI=1S/C21H35NO/c1-7-21(5,6)20-10-8-19(9-11-20)12-16(2)13-22-14-17(3)23-18(4)15-22/h8-11,16-18H,7,12-15H2,1-6H3/t16?,17-,18-/m0/s1
Formel vikt 317.2719