Läs mer
trans-Bifenthrin, TRC
Produktkod. 30134402
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134402 100 mg
30007089 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134402 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB383005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83322-02-5
Molekylformel C23H22ClF3O2
Molekylvikt (g/mol) 422.87
Synonym trans-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethylcyclopropanecarboxylic Acid (2-methyl[1,1'-biphenyl]-3-yl)methyl Ester;
IUPAC-namn (2-methyl-3-phenylphenyl)methyl 3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate
LEDER CC1=C(C=CC=C1COC(=O)[C@H]2[C@@H](C2(C)C)/C=C(/C(F)(F)F)\Cl)C3=CC=CC=C3
Specifikationer
Kemiskt namn eller material trans-Bifenthrin
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C23H22ClF3O2/c1-14-16(10-7-11-17(14)15-8-5-4-6-9-15)13-29-21(28)20-18(22(20,2)3)12-19(24)23(25,26)27/h4-12,18,20H,13H2,1-3H3/b19-12-
Procent renhet >95%