Läs mer
trans-Caffeic Acid, TRC
Produktkod. 30336774
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30336774 10 g
1 options
Produktkod. 30336774 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC07999010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans-Caffeic Acid
CAS 501-16-6
Molekylformel C9 H8 O4
Kvantitet 10 g
Synonym 2-Propenoic acid, 3-(3,4-dihydroxyphenyl)-, (2E)-, 2-Propenoic acid, 3-(3,4-dihydroxyphenyl)-, (E)-, Cinnamic acid, 3,4-dihydroxy-, (E)- (8CI), Cinnamic acid, 3,4-dihydroxy-, trans- (7CI), (2E)-(3,4-Dihydroxyphenyl)-2-propenoic acid, (2E)-3-(3,4-Dihydroxyphenyl)-2-propenoic acid, (E)-3,4-Dihydroxycinnamic acid, (E)-3-(3,4-Dihydroxyphenyl)-2-propenoic acid, (E)-3-(3,4-Dihydroxyphenyl)acrylic acid, (E)-Caffeic acid, 3,4-Dihydroxy-trans-cinnamic acid, trans-3,4-Dihydroxycinnamic acid, trans-Caffeic acid
InChI formel InChI=1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2+
LEDER OC(=O)\C=C\c1ccc(O)c(O)c1
IUPAC-namn (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
Molekylvikt (g/mol) 180.16
Formel vikt 180.0423