Läs mer
trans-Carboxy Glimepiride, TRC
Produktkod. 30367049
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367049 1 mg
30356888 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367049 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1784451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127554-90-9
Molekylformel C24H32N4O7S
Molekylvikt (g/mol) 520.6
Synonym trans-4-[[[[[4-[2-[[(3-Ethyl-2,5-dihydro-4-methyl-2-oxo-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]amino]carbonyl]amino]cyclohexanecarboxylic Acid
IUPAC-namn 4-[[4-[2-[(4-ethyl-3-methyl-5-oxo-2H-pyrrole-1-carbonyl)amino]ethyl]phenyl]sulfonylcarbamoylamino]cyclohexane-1-carboxylic acid
LEDER CCC1=C(CN(C1=O)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCC(CC3)C(=O)O)C
Specifikationer
Kemiskt namn eller material trans-Carboxy Glimepiride
Kvantitet 1 mg
InChI formel InChI=1S/C24H32N4O7S/c1-3-20-15(2)14-28(21(20)29)24(33)25-13-12-16-4-10-19(11-5-16)36(34,35)27-23(32)26-18-8-6-17(7-9-18)22(30)31/h4-5,10-11,17-18H,3,6-9,12-14H2,1-2H3,(H,25,33)(H,30,31)(H2,26,27,32)