Läs mer
(±)-trans-Chrysanthemic Acid, TRC
Produktkod. 30365416
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30365416 100 mg
1 options
Produktkod. 30365416 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC432790100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (±)-trans-Chrysanthemic Acid
CAS 705-16-8
Molekylformel C10H16O2
Kvantitet 100 mg
Synonym (1R,3R)-rel-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid (±)-trans-Chrysanthemumic Acid, (±)-trans-Chrysanthemummonocarboxylic Acid, DL-trans-Chrysanthemic Acid, dl-trans-Chrysanthemumic Acid, dl-trans-Chrysanthemummonocarboxylic Acid, trans-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid, trans-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid ester, trans-2,2-Dimethyl-3-(isobutenyl)cyclopropane-1-carboxylic Acid, trans-3,3-Dimethyl-2-(2-methyl-1-propenyl)cyclopropane-1-carboxylic Acid, trans-3,3-Dimethyl-2-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid, trans-Chrysanthemic Acid, trans-Chrysanthemum monocarboxylic Acid, trans-Chrysanthemumic Acid, (±)-trans-2,2-Dimethyl-3-isobutenyl-1-cyclopropanecarboxylic Acidtrans-(±)-2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylic Acid, trans-(±)-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid
InChI formel InChI=1S/C10H16O2/c1-6(2)5-7-8(9(11)12)10(7,3)4/h5,7-8H,1-4H3,(H,11,12)/t7-,8+/m1/s1
LEDER CC(=C[C@@H]1[C@@H](C(=O)O)C1(C)C)C
IUPAC-namn (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylic acid
Molekylvikt (g/mol) 168.23
Formel vikt 168.115