Läs mer
trans-Clomiphene Hydrochloride, TRC
Produktkod. 30007224
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30007224 5 mg -
30003746 2.5 mg 2.5mg
30006730 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007224 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5870355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14158-65-7
Molekylformel C26H29Cl2NO
Molekylvikt (g/mol) 442.42
Synonym (E)-2-[p-(2-chloro-1,2-diphenylvinyl)phenoxy]-triethylamine Hydrochloride, 2-[4-[(1E)-2-Chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethyl-ethanamine Hydrochloride, (E)-Clomiphene Hydrochloide, ICI 46476, Enclomifene Hydrochloride, Enclomiphene Hydrochloride, trans-Clomifene Hydrochloride;
IUPAC-namn 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;hydrochloride
LEDER CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.Cl
Specifikationer
Kemiskt namn eller material trans-Clomiphene Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C26H28ClNO.ClH/c1-3-28(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23;/h5-18H,3-4,19-20H2,1-2H3;1H/b26-25+;
Formel vikt 441.163
Procent renhet >95%