Läs mer
trans-Clopidogrel-MP Ethyl Ester Derivative(Mixture of Diastereomers), TRC
Produktkod. 30352572
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30352572 50 mg
1 options
Produktkod. 30352572 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC58727550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans-Clopidogrel-MP Ethyl Ester Derivative(Mixture of Diastereomers)
CAS 1331383-19-7
Molekylformel C27 H30 Cl N O6 S
Kvantitet 50 mg
Synonym Trans-Clopidogrel-Mp Ethyl Ester Derivative, (Mixture Of Diastereomers)
InChI formel InChI=1S/C27H30ClNO6S/c1-4-35-25(31)15-19-16-29(26(27(32)34-3)21-10-5-6-11-22(21)28)13-12-24(19)36-17-23(30)18-8-7-9-20(14-18)33-2/h5-11,14-15,24,26H,4,12-13,16-17H2,1-3H3/b19-15+
LEDER CCOC(=O)\C=C\1/CN(CCC1SCC(=O)c2cccc(OC)c2)C(C(=O)OC)c3ccccc3Cl
IUPAC-namn methyl 2-(2-chlorophenyl)-2-[(3E)-3-(2-ethoxy-2-oxoethylidene)-4-[2-(3-methoxyphenyl)-2-oxoethyl]sulfanylpiperidin-1-yl]acetate
Molekylvikt (g/mol) 532.048
Formel vikt 531.148