Läs mer
trans-Deltamethrin, TRC
Produktkod. 30368500
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30368500 25 mg
1 options
Produktkod. 30368500 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23050525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans-Deltamethrin
CAS 64363-96-8
Molekylformel C22H19Br2NO3
Kvantitet 25 mg
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, (S)-cyano(3-phenoxyphenyl)methyl ester, (1R,3S)-, Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, [1R-[1α(S*),3β]]-, NRDC 158B, RU 26979, trans-Deltamethrin, (1R,3S)-3-(2,2-Dibromoethenyl)-2,2-dimethylcyclopropanecarboxylic Acid (S)-Cyano(3-phenoxyphenyl)methyl Ester, [1R-[1α(S*),3β]-3-(2,2-Dibromoethenyl)-2,2-dimethylcyclopropanecarboxylic Acid (S)-Cyano(3-phenoxyphenyl)methyl Ester, NRDC 158B, RU 26979, Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, (S)-cyano(3-phenoxyphenyl)methyl ester, (1R,3S)-, Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, [1R-[1α(S*),3β]]-
InChI formel InChI=1S/C22H19Br2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18-,20+/m1/s1
LEDER CC1(C)[C@H](C=C(Br)Br)[C@H]1C(=O)O[C@H](C#N)c2cccc(Oc3ccccc3)c2
IUPAC-namn [(S)-cyano-(3-phenoxyphenyl)methyl] (1R,3S)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Molekylvikt (g/mol) 505.2
Formel vikt 502.973