Läs mer
trans-Hydroxy Glimepiride, TRC
Produktkod. 30408707
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30408707 10 mg
1 options
Produktkod. 30408707 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94253010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans-Hydroxy Glimepiride
CAS 600177-94-4
Molekylformel C24 H34 N4 O6 S
Kvantitet 10 mg
Synonym 1H-Pyrrole-1-carboxamide, 3-ethyl-2,5-dihydro-N-[2-[4-[[[[[trans-4-(hydroxymethyl)cyclohexyl]amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-4-methyl-2-oxo-, 3-Ethyl-2,5-dihydro-N-[2-[4-[[[[[trans-4-(hydroxymethyl)cyclohexyl]amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-4-methyl-2-oxo-1H-pyrrole-1-carboxamide, trans-Hydroxyglimepiride
InChI formel InChI=1S/C24H34N4O6S/c1-3-21-16(2)14-28(22(21)30)24(32)25-13-12-17-6-10-20(11-7-17)35(33,34)27-23(31)26-19-8-4-18(15-29)5-9-19/h6-7,10-11,18-19,29H,3-5,8-9,12-15H2,1-2H3,(H,25,32)(H2,26,27,31)/t18-,19-
LEDER CCC1=C(C)CN(C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)N[C@@H]3CC[C@@H](CO)CC3)C1=O
IUPAC-namn 4-ethyl-N-[2-[4-[[4-(hydroxymethyl)cyclohexyl]carbamoylsulfamoyl]phenyl]ethyl]-3-methyl-5-oxo-2H-pyrrole-1-carboxamide
Molekylvikt (g/mol) 506.62
Formel vikt 506.2199