Läs mer
trans-Linalool Oxide, TRC
Produktkod. 30479997
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479997 50 mg
30466987 10 mg
30466900 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479997 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46597550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34995-77-2
Molekylformel C10H18O2
Molekylvikt (g/mol) 170.25
Synonym trans-5-Ethenyltetrahydro-α,α,5-trimethyl-2-furanmethanol, Tetrahydro-α,α,5-trimethyl-5-vinyl-furfuryl Alcohol, rel-(2R,5R)-5-Ethenyltetrahydro-α,α,5-trimethyl-2-furanmethanol, (E)-Furanoid Linalool Oxide, (E)-Linalool Oxide, (E)-Linalool Oxide A, e-Linalool Oxide (furanoid), Epoxydihydrolinalool I, Linalool A Oxide, Linalool Oxide 2, Linalool Oxide A, Linalool Oxide II, trans-2-Methyl-2-vinyl-5-(1-hydroxy-1-methylethyl)tetrahydrofuran, trans-2-Vinyl-2-methyl-5-(1-hydroxy-1-methylethyl)tetrahydrofuran, trans-Epoxylinalool, trans-Furan Linalool Oxide, trans-Furanoid Linalool Oxide, trans-Linalool 3,6-oxide, trans-Linalool Oxide, trans-Linalool Oxide (furanoid), trans-f-Linalool Oxide, trans-α,α,5-Trimethyl-5-vinyltetrahydrofurfuryl Alcohol
IUPAC-namn 2-[(2R,5R)-5-ethenyl-5-methyloxolan-2-yl]propan-2-ol
LEDER C=C[C@]1(C)CC[C@H](C(C)(O)C)O1
Specifikationer
Kemiskt namn eller material trans-Linalool Oxide
Kvantitet 50 mg
InChI formel InChI=1S/C10H18O2/c1-5-10(4)7-6-8(12-10)9(2,3)11/h5,8,11H,1,6-7H2,2-4H3/t8-,10+/m1/s1
Formel vikt 170.1307