Learn More
trans-p-Methoxycinnamaldehyde, TRC
Product Code. 30471421
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
30471421 10 g 10g
1 options
Product Code. 30471421 Supplier Toronto Research Chemicals Supplier No. TRCM26213010G
 Restricted Item
Beräknad leverans: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material trans-p-Methoxycinnamaldehyde
CAS 24680-50-0
Molecular Formula C10 H10 O2
Quantity 10 g
Synonym 2-Propenal, 3-(4-methoxyphenyl)-, (2E)-, 2-Propenal, 3-(4-methoxyphenyl)-, (E)-, Cinnamaldehyde, p-methoxy-, (E)- (8CI), (2E)-3-(4-Methoxyphenyl)-2-propenal, (E)-3-(4-Methoxyphenyl)-2-propenal, (E)-3-(4-Methoxyphenyl)acrolein, (E)-3-(4-Methoxyphenyl)acrylaldehyde, (E)-3-(4-Methoxyphenyl)propenal, (E)-3-(4-methoxyphenyl)acrylaldehyde, (E)-4-Methoxycinnamaldehyde, (E)-4'-Methoxycinnamaldehyde, (E)-p-Methoxycinnamaldehyde, E-3-(4-Methoxyphenyl)acrylaldehyde, NSC 134287, trans-3-(4-Methoxyphenyl)acrolein, trans-3-(4-Methoxyphenyl)prop-2-enal, trans-4-Methoxycinnamaldehyde, trans-p-Methoxycinnamaldehyde
InChI Formula InChI=1S/C10H10O2/c1-12-10-6-4-9(5-7-10)3-2-8-11/h2-8H,1H3/b3-2+
SMILES COc1ccc(\C=C\C=O)cc1
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enal
Molecular Weight (g/mol) 162.19
Formula Weight 162.0681