Läs mer
(-)-trans-Phenothrin, TRC
Produktkod. 17973225
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17973225 500 mg
17961415 250 mg
17935025 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17973225 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP318108500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 26046-85-5
Molekylformel C23 H26 O3
Molekylvikt (g/mol) 350.45
Synonym D-trans-Phenothrin, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-, (3-phenoxyphenyl)methyl ester, (1R,3R)- (ACI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, (3-phenoxyphenyl)methyl ester, (1R,3R)- (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, (3-phenoxyphenyl)methyl ester, (1R-trans)- (ZCI), (-)-trans-Phenothrin, (1R-trans)-Biophenothrin, 1R,3R-Phenothrin, 1R-trans-Phenothrin, 3-Phenoxybenzyl (+)-trans-chrysanthemate
IUPAC-namn (3-phenoxyphenyl)methyl (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
LEDER CC(=C[C@@H]1[C@@H](C(=O)OCc2cccc(Oc3ccccc3)c2)C1(C)C)C
Specifikationer
Kemiskt namn eller material D-trans-Phenothrin
Kvantitet 500 mg
InChI formel InChI=1S/C23H26O3/c1-16(2)13-20-21(23(20,3)4)22(24)25-15-17-9-8-12-19(14-17)26-18-10-6-5-7-11-18/h5-14,20-21H,15H2,1-4H3/t20-,21+/m1/s1
Formel vikt 350.1882