Läs mer
trans R-138727, (Prasugrel Metabolite) -(>85%)(Mixture of Diastereomers), TRC
Produktkod. 30134454
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134454 10 mg
30133632 5 mg
17967174 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR07019510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 239466-74-1
Molekylformel C18 H20 F N O3 S
Molekylvikt (g/mol) 349.42
Synonym (2E)-2-[1-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4-mercapto-3-piperidinylidene]acetic Acid
IUPAC-namn (2E)-2-[1-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4-sulfanylpiperidin-3-ylidene]acetic acid
LEDER OC(=O)\C=C\1/CN(CCC1S)C(C(=O)C2CC2)c3ccccc3F
Specifikationer
Kemiskt namn eller material trans R-138727 (mixture of diastereomers)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C18H20FNO3S/c19-14-4-2-1-3-13(14)17(18(23)11-5-6-11)20-8-7-15(24)12(10-20)9-16(21)22/h1-4,9,11,15,17,24H,5-8,10H2,(H,21,22)/b12-9+
Formel vikt 349.1148
Procent renhet >95%