Läs mer
trans-Resveratrol 3-O-Beta-D-Glucuronide, TRC
Produktkod. 30137624
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137624 2.5 mg 2.5mg
30135223 5 mg -
30007766 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137624 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1500152.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 387372-17-0
Molekylformel C18 H18 O9
Molekylvikt (g/mol) 378.33
Synonym Trans-Resveratrol-B-O-B-D-Glucuronide
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[3-hydroxy-5-(4-hydroxyphenyl)phenoxy]tetrahydropyran-2-carboxylic acid
LEDER O[C@H]1[C@H](Oc2cc(O)cc(c2)c3ccc(O)cc3)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O
Specifikationer
Kemiskt namn eller material trans-Resveratrol-3-O-beta-D-Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C18H18O9/c19-10-3-1-8(2-4-10)9-5-11(20)7-12(6-9)26-18-15(23)13(21)14(22)16(27-18)17(24)25/h1-7,13-16,18-23H,(H,24,25)/t13-,14-,15+,16-,18+/m0/s1
Formel vikt 378.095
Procent renhet >95%