Läs mer
trans Resveratrol 3-Sulfate Sodium Salt, TRC
Produktkod. 30008605
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008605 10 mg
30006430 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR15004010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 858127-11-4
Molekylformel C14 H11 Na O6 S
Molekylvikt (g/mol) 330.29
Synonym 1,3-Benzenediol, 5-[(1E)-2-(4-hydroxyphenyl)ethenyl]-, 1-(hydrogen sulfate), sodium salt (1:1), 1,3-Benzenediol, 5-[(1E)-2-(4-hydroxyphenyl)ethenyl]-, 1-(hydrogen sulfate), monosodium salt (9CI), 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol 1-(hydrogen sulfate) monosodium salt, Resveratrol 3-O-sulfate monosodium salt, trans-Resveratrol 3-sulfate sodium salt
LEDER [Na+].Oc1ccc(\C=C\c2cc(O)cc(OS(=O)(=O)[O-])c2)cc1
Specifikationer
Kemiskt namn eller material trans Resveratrol 3-Sulfate sodium salt
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C14H12O6S.Na/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(8-11)20-21(17,18)19;/h1-9,15-16H,(H,17,18,19);/q;+1/p-1/b2-1+;
Formel vikt 330.0174
Procent renhet >95%