Läs mer
trans Resveratrol Penta-O-acetyl-3-Beta-D-glucuronide Methyl Ester, TRC
Produktkod. 30410153
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30410153 10 mg
1 options
Produktkod. 30410153 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR15003010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans Resveratrol Penta-O-acetyl-3-beta-D-glucuronide Methyl Ester
CAS 490028-22-3
Molekylformel C31H32O14
Kvantitet 10 mg
Synonym 3-(acetyloxy)-5-[(1E)-2-[4-(acetyloxy)phenyl]ethenyl]phenyl-beta-D-Glucopyranosiduronic Acid Methyl Ester Triacetate
InChI formel InChI=1S/C31H32O14/c1-16(32)39-23-11-9-21(10-12-23)7-8-22-13-24(40-17(2)33)15-25(14-22)44-31-29(43-20(5)36)27(42-19(4)35)26(41-18(3)34)28(45-31)30(37)38-6/h7-15,26-29,31H,1-6H3/b8-7+/t26-,27-,28-,29+,31+/m0/s1
LEDER COC(=O)[C@H]1O[C@@H](Oc2cc(OC(=O)C)cc(\C=C\c3ccc(OC(=O)C)cc3)c2)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-[3-acetyloxy-5-[(E)-2-(4-acetyloxyphenyl)ethenyl]phenoxy]oxane-2-carboxylate
Molekylvikt (g/mol) 628.58
Formel vikt 628.1792