Läs mer
trans Resveratrol Penta-O-acetyl-4’-Beta-D-glucuronide Methyl Ester, TRC
Produktkod. 30426109
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30426109 1 mg
1 options
Produktkod. 30426109 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1500351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans-Resveratrol Penta-O-acetyl-4'-beta-D-glucuronide Methyl Ester
CAS 490028-19-8
Molekylformel C31H32O14
Kvantitet 1 mg
Synonym 4-[(1E)-2-[3,5-Bis(acetyloxy)phenyl]ethenyl]phenyl-β-D-glucopyranosiduronic Acid Methyl Ester Triacetate, (E)-1-(3,5-Diacetoxyphenyl)-2-(4’-O-2,3,4-triacetyl
InChI formel InChI=1S/C31H32O14/c1-16(32)39-24-13-22(14-25(15-24)40-17(2)33)8-7-21-9-11-23(12-10-21)44-31-29(43-20(5)36)27(42-19(4)35)26(41-18(3)34)28(45-31)30(37)38-6/h7-15,26-29,31H,1-6H3/b8-7+/t26?,27?,28?,29?,31-/m1/s1
LEDER O=C([C@@H]1[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C=C2)=CC=C2/C=C/C3=CC(OC(C)=O)=CC(OC(C)=O)=C3)O1)OC
IUPAC-namn (2S,3R,4S,5S,6S)-2-(4-((E)-3,5-diacetoxystyryl)phenoxy)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
Molekylvikt (g/mol) 628.58
Formel vikt 628.18