Läs mer
trans-trans-10,11-Epoxy Farnesenic Acid Methyl Ester, TRC
Produktkod. 30004199
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004199 25 mg
30006293 10 mg
30008070 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004199 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE58940025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24198-95-6
Molekylformel C16 H26 O3
Molekylvikt (g/mol) 266.38
Synonym (2E,6E)- 9-(3,3-Dimethyl-2-oxiranyl)-3,7-dimethyl-2,6-nonadienoic Acid Methsyl Ester, (E,E)-(±)-10,11-Epoxy-3,7,11-trimethyl-2,6-dodecadienoic Acid Methyl Ester, (E,E)-Methyl 3,7,11-trimethyl-10,11-epoxy-2,6-dodecadienoate, (±)-Juvenile Hormone III, Methyl RS-(±)-10,11-Epoxyfarnesate;
IUPAC-namn methyl (2E,6E)-9-(3,3-dimethyloxiran-2-yl)-3,7-dimethylnona-2,6-dienoate
LEDER COC(=O)\C=C(/C)\CC\C=C(/C)\CCC1OC1(C)C
Specifikationer
Kemiskt namn eller material trans-trans-10,11-Epoxy Farnesenic Acid Methyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C16H26O3/c1-12(9-10-14-16(3,4)19-14)7-6-8-13(2)11-15(17)18-5/h7,11,14H,6,8-10H2,1-5H3/b12-7+,13-11+
Formel vikt 266.1882
Procent renhet >95%