Läs mer
(+)-trans-Verbenol, TRC
Produktkod. 30455435
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455435 25 mg
30453555 100 mg
30452089 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV12802025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22339-08-8
Molekylformel C10H16O
Molekylvikt (g/mol) 152.24
Synonym (1R,2S,5R)-4,6,6-Trimethyl-bicyclo[3.1.1]hept-3-en-2-ol, trans-(+)-2-Pinen-4-ol, [1R-(1alpha,2alpha,5alpha)]-4,6,6-Trimethyl-Bicyclo[3.1.1]hept-3-en-2-ol, (1R)-trans-Verbenol, (1R,4S,5R)-(+)-trans-Verbenol, (R)-trans-Verbenol, R-(+)-trans-Verbenol
IUPAC-namn (1R,2S,5R)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol
LEDER CC1=C[C@H](O)[C@@H]2C[C@H]1C2(C)C
Specifikationer
Kemiskt namn eller material (+)-trans-Verbenol
Kvantitet 25 mg
InChI formel InChI=1S/C10H16O/c1-6-4-9(11)8-5-7(6)10(8,2)3/h4,7-9,11H,5H2,1-3H3/t7-,8+,9+/m1/s1
Formel vikt 152.1201