missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Transferrin Receptor Monoclonal Antibody (3B82A1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA140198
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA1-40198 detects Transferrin receptor 1 from human samples. MA1-40198 has been successfully used in flow cytometry, ELISA, immunohistochemistry (frozen), immunoprecipitation, and Western blot applications. The MA1-40198 immunogen is human Transferrin receptor 1.
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
Specifikationer
| Transferrin Receptor | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| P02786 | |
| TFRC | |
| Human Transferrin receptor 1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 3B82A1 | |
| Unconjugated | |
| TFRC | |
| 2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr | |
| Mouse | |
| Protein G | |
| RUO | |
| 7037 | |
| 4°C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering