missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human ALK tyrosine kinase receptor / CD246 siRNA
Blocks transcription and translation of ALK tyrosine kinase receptor / CD246 from Human.
Brand: enQuireBio™ QX1-2nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_004295 | |
siRNA | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
CD246, NBLST3, Anaplastic lymphoma kinase | |
238 |
Lyophilized. | |
2 nmol | |
Human | |
ALK | |
SK-N-MC and NB-39-nu Neuroblastoma Cell Lines. |