Learn More
enQuireBio™ TranslationBlocker Human Caspase-1 siRNA
Blocks transcription and translation of Caspase-1 from Human.
Brand:  enQuireBio™ QX38-5nmol
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16059221
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
NP_001214
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
834
5 nmol
Human
Human Primary Keratinocytes.
CASP1
siRNA
Interleukin 1, Beta, Convertase, ICE, Caspase 1
Lyophilized