missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human CTLA4 siRNA
Blocks transcription and translation of CTLA4 from Human.
Brand: enQuireBio™ QX47-2nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_005205 | |
siRNA | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
Cytotoxic T-Lymphocyte Associated Protein 4, CTLA-3, CD152, Celiac3, Celiac Disease 3, Cytotoxic T-Lymphocyte-Associated Antigen 4 | |
1493 |
Lyophilized. | |
2 nmol | |
Human | |
CTLA4 | |
CLL Chronic Lymphoid Leukemia Cells. |