Läs mer
Kartell™ Transparent Polypropylene Powder Funnels
BIOPHARMA
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
Brand:  Kartell™ 0016700
567.15 SEK valid until 2026-04-30
BÄSTA PRIS på kampanj! Använd kampanjkod: "28507" för att få ditt kampanjpris.
Produktkod. 12539746
  • 597.00 SEK / Paket med 20
Tillbaka till labbet
Använd kampanjkoden 28507
Visa alla kampanjer
Bara null kvar
Beräknad leverans: 10-04-2026
Beskrivning
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
  • 60° angled sides
  • External ribbing avoids pressure lock
  • Can be used with liquids
  • Autoclavable