Läs mer
VITLAB™ Transparent Polypropylene Wide Mouth VITsafe™ Safety Wash Bottles
Made from polypropylene. Vitlab™ Transparent Polypropylene Wide Mouth VITsafe™ Safety Wash Bottles offer a precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.
Brand: VITLAB™ 1451829
Beskrivning
- Polypropylene spray tube
- With coloured VENT-CAP for easy visual identification
- Safety imprint in accordance to Directive (EC) No. 1272/2008 (GHS):
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- Practically no leakage or dripping with the VENT-CAP