Läs mer
VITLAB™ Transparent Polypropylene Wide Mouth VITsafe™ Safety Wash Bottles
Made from polypropylene. Vitlab™ Transparent Polypropylene Wide Mouth VITsafe™ Safety Wash Bottles offer a precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.
Brand:  VITLAB™ 1451829
Produktkod. 10634883
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
  • Polypropylene spray tube
  • With coloured VENT-CAP for easy visual identification
  • Safety imprint in accordance to Directive (EC) No. 1272/2008 (GHS):
    • Material name in German, English, French and Spanish
    • Chemical formula, CAS No, hazard pictogram, signal word
    • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
  • Practically no leakage or dripping with the VENT-CAP